Proteintech
Manufacturer Code:170731AP150UL
Catalog # 50-173-1368
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LZTFL1 Fusion Protein Ag10919 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: leucine zipper transcription factor-like 1; Leucine zipper transcription factor-like protein 1; LZTFL1
Gene Aliases: 5530402H04Rik; 6130400H19Rik; AI414725; AW048545; BBS17; LZTFL1
UniProt ID: (Human) Q9NQ48, (Mouse) Q9JHQ5, (Rat) Q562C6
Entrez Gene ID: (Human) 54585, (Mouse) 93730, (Rat) 316102
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.