Invitrogen
Manufacturer Code:PA576098
Catalog # PIPA576098
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human LZIP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic leucine zipper protein; cAMP-responsive element-binding protein 3; CREB-3; cyclic AMP response element (CRE)-binding protein/activating transcription factor 1; Cyclic AMP-responsive element-binding protein 3; Cyclic AMP-responsive element-binding protein 3 CREB-3 cAMP-responsive element-binding protein 3 Leucine zipper protein Luman Transcription factor LZIP-alpha CREB3 LZIP; leucin zipper proitein; Leucine zipper protein; Luman; N-terminal Luman; Processed cyclic AMP-responsive element-binding protein 3; small leucine zipper protein; transcription factor LZIP; Transcription factor LZIP-alpha
Gene Aliases: AU044960; AW538053; C80076; CREB3; LUMAN; LZIP; LZIP-1; LZIP-2; sLZIP
UniProt ID: (Human) O43889
Entrez Gene ID: (Human) 10488, (Mouse) 12913
Molecular Function: CREB transcription factor basic leucine zipper transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.