Invitrogen
Manufacturer Code:PA5103568
Catalog # PIPA5103568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human LYPLA2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-protein thioesterase 2; Acyl-protein thioesterase 2 APT2 APT-2 DJ886K2.4 LPL-II LYPLA2 LysoII Lysophospholipase 2 lysophospholipase II lysoPLA II mLyso II testicular tissue protein Li 3; APT-2; LPL-II; Lysophospholipase 2; Lysophospholipase II; lysoPLA II; mLyso II; testicular tissue protein Li 3
Gene Aliases: APT-2; APT2; DJ886K2.4; LYPLA2; LysoII
UniProt ID: (Human) Q7Z4Z2, (Rat) Q9QYL8, (Mouse) Q9WTL7
Entrez Gene ID: (Human) 11313, (Rat) 83510, (Mouse) 26394
Molecular Function: hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.