Novus Biologicals
Manufacturer Code:NBP30472920UL
Catalog # NB094453
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 147-231 of human LYPLA2 (NP_009191.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-protein thioesterase 2; acyl-protein thioesterase 2 APT2 DJ886K2.4 EC 3.1.2 EC 3.1.2.- LPL-II lysophospholipase IIAPT-2LysoPLA II; APT-2; LPL-II; Lysophospholipase II; lysoPLA II; testicular tissue protein Li 3
Gene Aliases: APT-2; APT2; DJ886K2.4; LYPLA2
UniProt ID: (Human) Q7Z4Z2
Entrez Gene ID: (Human) 11313
Molecular Function: hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.