Invitrogen
Manufacturer Code:MA534855
Catalog # PIMA534855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JE47-60 |
Immunogen | Recombinant protein within Lysophospholipase 1 aa 100-250. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 25KDL Acyl-protein thioesterase 1 Apt1 APT-1 BOS_13842 CaIPLA2 Calcium-independent phospholipase A2 hAPT1 LPL1 LPL-I Lypla1 Lysophospholipase 1 Lysophospholipase I LYSOPLA Pla1a zgc:110260; Acyl-protein thioesterase 1; APT-1; LPL-I; Lysophospholipase 1; Lysophospholipase I; lysophospholipid-specific lysophospholipase; lysoPLA I
Gene Aliases: APT-1; APT1; hAPT1; LPL-I; LPL1; LYPLA1
UniProt ID: (Human) O75608
Entrez Gene ID: (Human) 10434
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.