Novus Biologicals
Manufacturer Code:NBP214209
Catalog # NBP214209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: LTTQQKLAGVTALNCWLPLWASFPQGPIGGANRDISILQCHGDCDPLV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-protein thioesterase 1; acyl-protein thioesterase-1 APT1 APT-1 EC 3.1.2 EC 3.1.2.- hAPT1 LPL1acyl-protein thioesterase 1 LPL-I Lysophospholipase 1 lysophospholipase ILYSOPLA lysophospholipid-specific lysophospholipase LysoPLA I; APT-1; LPL-I; Lysophospholipase 1; Lysophospholipase I; lysophospholipid-specific lysophospholipase; lysoPLA I
Gene Aliases: APT-1; APT1; hAPT1; LPL-I; LPL1; LYPLA1
UniProt ID: (Human) O75608
Entrez Gene ID: (Human) 10434
Molecular Function: hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.