Proteintech
Manufacturer Code:160551AP150UL
Catalog # 50-173-0681
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LYPLA1 Fusion Protein Ag9022 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl protein thioesterase 1 APT 1 APT1 hAPT1 LPL I LPL1 LYPLA1 Lysophospholipase 1 lysophospholipase I LYSOPLA LysoPLA I; Acyl-protein thioesterase 1; APT-1; LPL-I; lysophopholipase 1; Lysophospholipase 1; Lysophospholipase I; lysophospholipid-specific lysophospholipase; lysoPLA I; phospholipase 1a
Gene Aliases: 25KDL; APT-1; APT1; hAPT1; LPL-I; LPL1; LYPLA1; Pla1a
UniProt ID: (Human) O75608, (Rat) P70470, (Mouse) P97823
Entrez Gene ID: (Human) 10434, (Rat) 25514, (Mouse) 18777
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.