Proteintech
Manufacturer Code:133731AP150UL
Catalog # 50-172-8689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LYNX1 Fusion Protein Ag4195 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Endogenous prototoxin LYNX1; GC26; Ly-6 neurotoxin-like protein 1; Ly-6/neurotoxin-like protein 1; Ly6/neurotoxin 1 LYNX1 SLURP 2 SLURP2; secreted Ly-6/uPAR domain-containing protein 2; secreted Ly-6/uPAR-related protein 2; secreted Ly6/uPAR related protein 2; SLURP-2; Testicular tissue protein Li 112
Gene Aliases: AI838844; LYNX1; SLURP-2; SLURP2
UniProt ID: (Human) P0DP58, (Mouse) P0DP60
Entrez Gene ID: (Human) 66004, (Mouse) 23936, (Rat) 300018
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.