Abnova
Manufacturer Code:PAB3431
Catalog # 89-131-013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human LYN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of LYN.
For Research Use Only
Protein Aliases: FLJ26625; JTK8; Lck/Yes-related novel protein tyrosine kinase; p53Lyn; p56Lyn; Tyrosine-protein kinase Lyn; V-yes-1 Yamaguchi sarcoma viral related oncogene homolog
Gene Aliases: JTK8; LYN; p53Lyn; p56Lyn
UniProt ID: (Human) P07948
Entrez Gene ID: (Human) 4067
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.