Novus Biologicals
Manufacturer Code:NBP2852390.1ML
Catalog # NB033168
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human LYG2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.2.1.- LYGHMGC119046 lysozyme G-like 2 lysozyme g-like protein 2 MGC119047 MGC119049; lysozyme G-like 2; Lysozyme g-like protein 2
Gene Aliases: LYG2; LYGH; LYSG2
UniProt ID: (Human) Q86SG7
Entrez Gene ID: (Human) 254773
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.