R&D Systems
Manufacturer Code:MAB83291100
Catalog # MAB83291100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2489B |
Immunogen | Human embryonic kidney cell HEK293-derived human Lysosomal alpha-Glucosidase Ala70-Cys952 Accession &num P10253 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 70 kDa lysosomal alpha-glucosidase; 76 kDa lysosomal alpha-glucosidase; Acid alpha-Glucosidase Acid maltase Aglucosidase alfa EC 3.2.1.20 GAA glucosidase alpha acid LYAG lysosomal alpha-glucosidase Lysosomal alphaGlucosidase; Acid maltase; Aglucosidase alfa; Lysosomal alpha-glucosidase
Gene Aliases: GAA; LYAG
UniProt ID: (Human) P10253
Entrez Gene ID: (Human) 2548
Molecular Function:
glucosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.