Abnova
Manufacturer Code:H00008581B02
Catalog # 89-006-597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | LY6D (NP_003686.1 1 a.a. ~ 128 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human LY6D protein.
For Research Use Only
Protein Aliases: E48; E48 antigen; Ly-6D; Lymphocyte antigen 6D
Gene Aliases: E48; Ly-6D; LY6D
UniProt ID: (Human) Q14210
Entrez Gene ID: (Human) 8581
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.