Proteintech
Manufacturer Code:173611AP150UL
Catalog # 50-173-1530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LY6D Fusion Protein Ag10714 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E48 antigen; E48 E48 antigen Ly 6D LY6D Lymphocyte antigen 6D; Ly-6D; Lymphocyte antigen 6D; ThB; Thymocyte B-cell antigen
Gene Aliases: E48; Ly-61; Ly-6D; Ly61; LY6D; Thb
UniProt ID: (Human) Q14210, (Mouse) P35459
Entrez Gene ID: (Human) 8581, (Mouse) 17068
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.