Novus Biologicals
Manufacturer Code:NB10074457
Catalog # NB10074457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues R(113) Y A C R G S G T C Q M D A(126) of rat LXR beta. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: liver X nuclear receptor beta; Liver X receptor beta; Liver X receptor beta LXR-b NER-I NERliver X receptor-beta nuclear orphan receptor LXR-beta Nuclear receptor NER Nuclear receptor subfamily 1 group H member 2 nuclear receptor subfamily 1 group H member 2 oxysterols receptor LXR-beta RIP15 steroid hormone-nuclear receptor NER Ubiquitously-expressed nuclear receptorLXRB UNRLX receptor beta; LX receptor beta; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; Oxysterols receptor LXR-beta; steroid hormone-nuclear receptor NER; Ubiquitously-expressed nuclear receptor
Gene Aliases: LXR-b; LXRB; NER; NER-I; NR1H2; RIP15; UNR
UniProt ID: (Human) E7EWA6
Entrez Gene ID: (Human) 7376
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.