Invitrogen
Manufacturer Code:711097
Catalog # PI711097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 8 HCLC |
Immunogen | Peptides corresponding to Human NR1H2 (aa 244-257 57-74) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles.,store at -20°C |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: liver X nuclear receptor beta; Liver X receptor beta; LX receptor beta; LXR beta; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; Oxysterols receptor LXR-beta; Retinoid X receptor-interacting protein No.15; steroid hormone-nuclear receptor NER; Ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 2
Gene Aliases: AI194859; LXR; LXR-b; LXRB; LXRbeta; NER; NER-I; NER1; NR1H2; OR-1; RIP15; UNR; Unr2; UR
UniProt ID: (Human) P55055, (Mouse) Q60644
Entrez Gene ID: (Human) 7376, (Mouse) 22260
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.