Invitrogen
Manufacturer Code:PA569243
Catalog # PIPA569243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat LXR beta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: liver X nuclear receptor beta; Liver X receptor beta; LX receptor beta; LXR beta; NR1H2 LXR-b NER UNR NER-I RIP15 nuclear receptor subfamily 1 group H member 2 ubiquitously-expressed nuclear receptor LXRB LX receptor beta liver X receptor beta liver X receptor-beta nuclear orphan receptor LXR-beta oxysterols receptor LXR-beta steroid hormone-nuclear receptor NER; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; Orphan nuclear receptor OR-1; Oxysterols receptor LXR-beta; Retinoid X receptor-interacting protein No.15; steroid hormone-nuclear receptor NER; ubiquitous receptor; Ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 2; UR
Gene Aliases: AI194859; LXR; LXR-b; LXRB; LXRbeta; NER; NER-I; NER1; NR1H2; OR-1; RIP15; UNR; Unr2; UR
UniProt ID: (Human) E7EWA6, (Bovine) Q5BIS6, (Mouse) Q60644, (Rat) Q62755
Entrez Gene ID: (Human) 7376, (Horse) 100052399, (Pig) 100525290, (Rabbit) 100342868, (Bovine) 509622, (Dog) 610057, (Mouse) 22260, (Rat) 58851, (Guinea pig) 100719951
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.