Invitrogen
Manufacturer Code:PA529771
Catalog # PIPA529771
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 162 and 325 of Human LXR beta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: liver X nuclear receptor beta; Liver X receptor beta; LX receptor beta; LXR beta; LXR-b LXRB NER NER-I RIP15 UNR LX receptor beta liver X nuclear receptor beta liver X receptor beta liver X receptor-beta nuclear orphan receptor LXR-beta nuclear receptor NER oxysterols receptor LXR-beta steroid hormone-nuclear receptor NER ubiquitously-expressed nuclear receptor; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; Oxysterols receptor LXR-beta; Retinoid X receptor-interacting protein No.15; steroid hormone-nuclear receptor NER; Ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 2
Gene Aliases: AI194859; LXR; LXR-b; LXRB; LXRbeta; NER; NER-I; NER1; NR1H2; OR-1; RIP15; UNR; Unr2; UR
UniProt ID: (Human) E7EWA6, (Mouse) Q60644
Entrez Gene ID: (Human) 7376, (Mouse) 22260
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.