Invitrogen
Manufacturer Code:PA1331B
Catalog # PIPA1331B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues C L(432) R L Q D K K L P P L L S E(445) of human LXR alpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: liver X nuclear receptor alpha variant 1; liver X nuclear receptor beta; Liver X Receptor; Liver X receptor alpha; Liver X receptor beta; LX receptor beta; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 2; nuclear receptor subfamily 1, group H, member 3; Oxysterols receptor LXR-alpha; Oxysterols receptor LXR-beta; steroid hormone-nuclear receptor NER; Ubiquitously-expressed nuclear receptor
Gene Aliases: LXR-a; LXR-b; LXRA; LXRB; NER; NER-I; NR1H2; NR1H3; RIP15; RLD-1; UNR
UniProt ID: (Human) E7EWA6, (Human) Q13133
Entrez Gene ID: (Human) 7376, (Human) 10062
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.