Novus Biologicals
Manufacturer Code:NB1001465
Catalog # NB1001465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence CRLQDKKLPPLLSEI corresponding to internal region according to NP_005684.2 NP_009052.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: liver X nuclear receptor alpha variant 1; Liver X receptor alpha; LXR-a LXRA NR1H3 nuclear receptor subfamily 1 group H member 3 RLD-1; Nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 3; Oxysterols receptor LXR-alpha
Gene Aliases: LXR-a; LXRA; NR1H3; RLD-1
UniProt ID: (Human) Q13133
Entrez Gene ID: (Human) 10062
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.