Invitrogen
Manufacturer Code:PA19066
Catalog # 01-674-650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Internal sequence corresponding to amino acids CRLQDKKLPPLLSEI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: liver X nuclear receptor alpha variant 1; liver X nuclear receptor beta; Liver X receptor alpha; Liver X receptor beta; LX receptor beta; LXR alpha; LXR beta; NR1H3 LXRA LXR-a RLD-1 nuclear receptor subfamily 1 group H member 3 liver X receptor alpha NR1H2 NER UNR LXR-b NER-I RIP15 nuclear rec; nuclear orphan receptor LXR-alpha; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 2; nuclear receptor subfamily 1, group H, member 3; Orphan nuclear receptor OR-1; Oxysterols receptor LXR-alpha; Oxysterols receptor LXR-beta; Retinoid X receptor-interacting protein No.15; RLD-1; steroid hormone-nuclear receptor NER; ubiquitous receptor; Ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 1; ubiquitously-expressed nuclear receptor 2; UR
Gene Aliases: AI194859; AU018371; LXR; LXR-a; LXR-b; LXRA; LXRalpha; LXRB; LXRbeta; NER; NER-I; NER1; NR1H2; NR1H3; OR-1; RIP15; RLD-1; RLD1; UNR; Unr1; Unr2; UR
UniProt ID: (Human) E7EWA6, (Human) Q13133, (Mouse) Q60644, (Rat) Q62755, (Mouse) Q9Z0Y9
Entrez Gene ID: (Human) 7376, (Human) 10062, (Mouse) 22260, (Rat) 58851, (Mouse) 22259, (Rat) 58852
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.