Novus Biologicals
Manufacturer Code:NBP272550H
Catalog # NBP272550H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A5 |
Immunogen | Recombinant protein fragment corresponding to amino acids 69-329 of human LXR alpha/NR1H3 (NP_005684) produced in E.coli. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: alpha Liver X receptor alpha liver X receptor-alpha LXRA Nuclear receptor subfamily 1 group H member 3 nuclear receptor subfamily 1 group H member 3 oxysterols receptor LXR-alpha RLD-1; liver X nuclear receptor alpha variant 1; Liver X receptor alpha; Nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 3; Oxysterols receptor LXR-alpha
Gene Aliases: LXR-a; LXRA; NR1H3; RLD-1
UniProt ID: (Human) Q13133
Entrez Gene ID: (Human) 10062
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.