Invitrogen
Manufacturer Code:PA116824
Catalog # APPA6824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of the human LXR protein sequence (between residues 50-150). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: liver X nuclear receptor alpha variant 1; Liver X receptor alpha; Liver X receptor alpha Liver X receptor beta Nuclear receptor NER Ubiquitously expressed nuclear receptor LXRa LXRb NER NERI NR1H2 NR1H3; LXR alpha; nuclear oxysterol receptor LxR-alpha; Nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 3; Oxysterols receptor LXR-alpha; ubiquitously-expressed nuclear receptor 1
Gene Aliases: AU018371; LXR; LXR-a; LXRA; NR1H3; RLD-1; RLD1; Unr1
UniProt ID: (Human) Q13133, (Mouse) Q9Z0Y9
Entrez Gene ID: (Pig) 397553, (Human) 10062, (Mouse) 22259
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.