Invitrogen
Manufacturer Code:PA566014
Catalog # PIPA566014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human LUC7L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LUC7-like; LUC7L1; Putative RNA-binding protein Luc7-like 1; Putative SR protein LUC7B1; sarcoplasmic reticulum protein LUC7B1; SR+89
Gene Aliases: hLuc7B1; Luc7; LUC7B1; LUC7L; LUC7L1; SR+89
UniProt ID: (Human) Q9NQ29
Entrez Gene ID: (Human) 55692
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.