Proteintech
Manufacturer Code:170851AP150UL
Catalog # 50-173-1376
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LUC7L Fusion Protein Ag9170 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LUC7 LIKE LUC7 like (S. cerevisiae) LUC7B1 LUC7L LUC7L1 Putative SR protein LUC7B1 SR+89; LUC7-like; Putative RNA-binding protein Luc7-like 1; Putative SR protein LUC7B1; sarcoplasmic reticulum protein LUC7B1; SR+89
Gene Aliases: 1810045C04Rik; 2410018D03Rik; hLuc7B1; Luc7; LUC7B1; LUC7L; LUC7L1; SR+89
UniProt ID: (Human) Q9NQ29, (Mouse) Q9CYI4
Entrez Gene ID: (Human) 55692, (Rat) 360503, (Mouse) 66978
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.