Abnova
Manufacturer Code:MAB20434
Catalog # 89-336-834
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ABOI-12 |
Immunogen | A synthetic peptide corresponding to human LTF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C for one year. After reconstitution at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody.
Protein Aliases: epididymis luminal protein 110; GIG12 HLF2 LF; Growth-inhibiting protein 12; Kaliocin-1; lactoferricin; Lactoferricin-H; Lactoferrin; lactoferroxin; Lactoferroxin-A; Lactoferroxin-B; Lactoferroxin-C; Lactotransferrin; Lfcin-H; neutrophil lactoferrin; Talalactoferrin
Gene Aliases: GIG12; HEL110; HLF2; LF; LTF
UniProt ID: (Human) P02788
Entrez Gene ID: (Human) 4057
Molecular Function: hydrolase protease serine protease transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.