Abnova
Manufacturer Code:MAB0524
Catalog # 89-107-968
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B8 |
Immunogen | Native purified human LTF. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse monoclonal antibody raised against native LTF.
For Research Use Only
Protein Aliases: epididymis luminal protein 110; GIG12; Growth-inhibiting protein 12; HLF2; Kaliocin-1; lactoferricin; Lactoferricin-H; Lactoferrin; lactoferroxin; Lactoferroxin-A; Lactoferroxin-B; Lactoferroxin-C; Lactotransferrin; LF; Lfcin-H; neutrophil lactoferrin; Talalactoferrin
Gene Aliases: GIG12; HEL110; HLF2; LF; LTF
UniProt ID: (Human) P02788
Entrez Gene ID: (Human) 4057
Molecular Function: hydrolase protease serine protease transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.