Novus Biologicals
Manufacturer Code:NBP30394920UL
Catalog # NB095702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 50 to the C-terminus of human LTC4S (NP_665874.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 4.4.1.20 leukotriene C4 synthase Leukotriene-C(4) synthase LTC4 synthase MGC33147; Glutathione S-transferase LTC4; Leukotriene C4 synthase; Leukotriene-C(4) synthase; Leukotriene-C4 synthase; LTC4 synthase
Gene Aliases: LTC4S
UniProt ID: (Human) Q16873
Entrez Gene ID: (Human) 4056
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.