Invitrogen
Manufacturer Code:PA585149
Catalog # PIPA585149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human LTBP4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ46318 antibody FLJ90018 antibody LTBP-4 antibody LTBP4L antibody LTBP4S antibody LTBP4 antibody latent-transforming growth factor beta-binding protein 4 antibody latent transforming growth factor-beta binding protein 4L antibody latent transforming growth factor beta binding protein 4 antibody; latent transforming growth factor-beta binding protein 4L; Latent-transforming growth factor beta-binding protein 4; LTBP-4
Gene Aliases: 2310046A13Rik; ARCL1C; LTBP-4; LTBP4; LTBP4L; LTBP4S
UniProt ID: (Human) Q8N2S1, (Mouse) Q8K4G1
Entrez Gene ID: (Human) 8425, (Mouse) 108075
Molecular Function:
annexin
calcium-binding protein
calmodulin
cell adhesion molecule
extracellular matrix glycoprotein
extracellular matrix protein
extracellular matrix structural protein
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.