Invitrogen
Manufacturer Code:PA599878
Catalog # PIPA599878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human LTBP1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9430031G15Rik 9830146M04 b2b1000Clo LanC (bacterial lantibiotic synthetase component C)-like 1 latent TGF beta binding protein latent transforming growth factor beta binding protein 1 latent-transforming growth factor beta-binding protein 1 Ltbp1 LTBP-1 MGC163161 Tgfb TGF-beta-1-BP-1 TGF-beta1-BP-1 Transforming growth factor beta-1-binding protein 1 transforming growth factor beta-1-masking protein large subunit transforming growth factor-beta (TGF-beta) masking protein large subunit; LanC (bacterial lantibiotic synthetase component C)-like 1; latent TGF beta binding protein; Latent-transforming growth factor beta-binding protein 1; LTBP-1; TGF-beta-1-BP-1; TGF-beta1-BP-1; Transforming growth factor beta-1-binding protein 1; Transforming growth factor beta-1-masking protein large subunit; transforming growth factor-beta (TGF-beta) masking protein large subunit
Gene Aliases: 9430031G15Rik; 9830146M04; b2b1000Clo; Ltbp-1; LTBP1; Tgfb
UniProt ID: (Human) Q14766, (Mouse) Q8CG19, (Rat) Q00918
Entrez Gene ID: (Human) 4052, (Mouse) 268977, (Rat) 59107
Molecular Function:
annexin
calcium-binding protein
calmodulin
cell adhesion molecule
extracellular matrix glycoprotein
extracellular matrix protein
extracellular matrix structural protein
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.