Invitrogen
Manufacturer Code:PA545075
Catalog # PIPA545075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human LTBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LanC (bacterial lantibiotic synthetase component C)-like 1; latent TGF beta binding protein; Latent transforming growth factor beta binding protein 1 MGC163161; latent transforming growth factor binding protein 1; Latent-transforming growth factor beta-binding protein 1; LTBP-1; LTBP1; TGF-beta-1-BP-1; TGF-beta1-BP-1; Transforming growth factor beta-1-binding protein 1; Transforming growth factor beta-1-masking protein large subunit; transforming growth factor-beta (TGF-beta) masking protein large subunit
Gene Aliases: 9430031G15Rik; 9830146M04; b2b1000Clo; Ltbp-1; LTBP1; Tgfb
UniProt ID: (Human) Q14766, (Mouse) Q8CG19, (Rat) Q00918
Entrez Gene ID: (Dog) 475720, (Bovine) 510559, (Human) 4052, (Rabbit) 100342431, (Horse) 100070522, (Mouse) 268977, (Guinea pig) 100733334, (Rat) 59107
Molecular Function: annexin calcium-binding protein calmodulin cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein extracellular matrix structural protein intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.