OriGene
Manufacturer Code:TA700155
Catalog # 50-167-7684
Quantity:
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8F4 |
Immunogen | Full length human recombit protein of human LTA4H produced in HEK293T cell. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Leukotriene A(4) hydrolase; Leukotriene A-4 hydrolase; leukotriene A4 hydrolase; LTA-4 hydrolase; testicular secretory protein Li 27; Tripeptide aminopeptidase LTA4H
Gene Aliases: LTA4; LTA4H
UniProt ID: (Human) P09960
Entrez Gene ID: (Human) 4048
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.