Proteintech
Manufacturer Code:137151AP150UL
Catalog # 50-172-8954
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LSS Fusion Protein Ag4655 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2,3-epoxysqualene--lanosterol cyclase; 2,3-epoxysqualene-lanosterol cyclase; 2,3-oxidosqualene: lanosterol cyclase; hOSC; hOSC Lanosterol synthase LSS OSC; Lanosterol synthase; OSC; Oxidosqualene--lanosterol cyclase
Gene Aliases: 2810025N20Rik; CTRCT44; D10Ertd116e; LSS; OSC
UniProt ID: (Human) P48449, (Mouse) Q8BLN5, (Rat) P48450
Entrez Gene ID: (Human) 4047, (Mouse) 16987, (Rat) 81681
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.