Proteintech
Manufacturer Code:101341AP150UL
Catalog # 50-172-6049
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LSM8 Fusion Protein Ag0178 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LSM8 homolog, U6 small nuclear RNA associated; LSM8 NAA38 YJR022W; LSM8 U6 small nuclear RNA associated; MAK31-like protein; N-alpha-acetyltransferase 38, NatC auxiliary subunit; U6 snRNA-associated Sm-like protein LSm8
Gene Aliases: 2010003I05Rik; AW214405; LSM8; NAA38
UniProt ID: (Human) O95777, (Mouse) Q6ZWM4
Entrez Gene ID: (Human) 51691, (Mouse) 76522, (Rat) 296913
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.