Invitrogen
Manufacturer Code:PA580781
Catalog # PIPA580781
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human LSM1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer-associated Sm protein; Cancer-associated Sm-like; cancer-associated Sm-like protein; cancer‐associated Sm protein; CASM YJL124C; LSM1 homolog, U6 small nuclear RNA associated; LSM1 mRNA degradation associated; LSM1, U6 small nuclear RNA associated; LSM1-like protein U6 small nuclear RNA associated; Small nuclear ribonuclear CaSm; U6 snRNA-associated Sm-like protein LSm1
Gene Aliases: CASM; LSM1; YJL124C
UniProt ID: (Human) O15116
Entrez Gene ID: (Human) 27257
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.