Invitrogen
Manufacturer Code:PA558069
Catalog # PIPA558069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human LSG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hLsg1; hLsg1 large subunit GTPase 1 homolog; Large subunit GTPase 1 homolog
Gene Aliases: 5830465I20; AA409273; D16Bwg1547e; LSG1; RGD1309089
UniProt ID: (Human) Q9H089, (Rat) Q5BJT6, (Mouse) Q3UM18
Entrez Gene ID: (Human) 55341, (Rat) 288029, (Mouse) 224092
Molecular Function:
G-protein
enzyme modulator
hydrolase
nucleic acid binding
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.