Novus Biologicals
Manufacturer Code:NBP287751
Catalog # NB035373
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human LSG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.6.1 EC 3.6.1.- FLJ11301 FLJ27294 hLsg1 large subunit GTPase 1 homolog large subunit GTPase 1 homolog (S. cerevisiae); hLsg1; Large subunit GTPase 1 homolog
Gene Aliases: LSG1
UniProt ID: (Human) Q9H089
Entrez Gene ID: (Human) 55341
Molecular Function: G-protein enzyme modulator hydrolase nucleic acid binding signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.