R&D Systems
Manufacturer Code:MAB2947100
Catalog # MAB2947100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 845404 |
Immunogen | NS0 mouse myeloma cell line transfected with LSECtin/CLEC4G Ser54-Cys293 Accession # Q6UXB4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-type lectin domain family 4 member G; C-type lectin domain family 4, member G; C-type lectin superfamily 4, member G; CLEC4G C-type lectin domain family 4 member G C-type lectin domain family 4 member G C-type lectin superfamily 4 member G DTTR431 liver and lymph node sinusoidal endothelial cell C-type lectin LP2698 LSECtin Q6UXB4 UNQ431; Liver and lymph node sinusoidal endothelial cell C-type lectin; LSECtin
Gene Aliases: CLEC4G; DTTR431; LP2698; LSECtin; UNQ431; UNQ431/PRO792
UniProt ID: (Human) Q6UXB4
Entrez Gene ID: (Human) 339390
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.