Invitrogen
Manufacturer Code:PA585426
Catalog # PIPA585426
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human LSD1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-dimethyl-L-lysine(4) FAD-dependent demethylase 1A; amine oxidase (flavin containing) domain 2; AOF2 antibody BHC110 antibody KDM1 antibody KIAA0601 antibody LSD1 antibody KDM1A antibody BRAF35-HDAC complex protein BHC110 antibody flavin-containing amine oxidase domain-containing protein 2 antibody FAD-binding protein BRAF35-HDAC complex 110 kDa subunit antibody lysine (K)-specific demethylase 1 antibody lysine-specific histone demethylase 1A antibody lysine-specific histone demethylase 1 antibody amine oxidase (flavin containing) domain 2 antibody lysine (K)-specific demethylase 1A antibody; BRAF35-HDAC complex protein BHC110; FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit; Flavin-containing amine oxidase domain-containing protein 2; lysine (K)-specific demethylase 1; lysine (K)-specific demethylase 1A; lysine-specific histone demethylase 1; Lysine-specific histone demethylase 1A; neuroprotective protein 3
Gene Aliases: AOF2; BHC110; CPRF; KDM1; KDM1A; KIAA0601; LSD1; RGD1562975
UniProt ID: (Human) Q8NDP6
Entrez Gene ID: (Human) 23028, (Rat) 500569
Molecular Function:
DNA binding protein
DNA methyltransferase
methyltransferase
nucleic acid binding
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.