Novus Biologicals
Manufacturer Code:NB10056496SS
Catalog # NB10056496T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 800-850 of human LSD1 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-dimethyl-L-lysine(4) FAD-dependent demethylase 1A; amine oxidase (flavin containing) domain 2; amine oxidase (flavin containing) domain 2 AOF2lysine-specific histone demethylase 1 BHC110FAD-binding protein BRAF35-HDAC complex 110 kDa subunit Flavin-containing amine oxidase domain-containing protein 2 KIAA0601KDM1 LSD1BRAF35-HDAC complex protein BHC110 lysine (K)-specific demethylase 1 lysine (K)-specific demethylase 1A lysine-specific histone demethylase 1A; BRAF35-HDAC complex protein BHC110; FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit; Flavin-containing amine oxidase domain-containing protein 2; lysine (K)-specific demethylase 1A; lysine-specific histone demethylase 1; Lysine-specific histone demethylase 1A
Gene Aliases: AOF2; BHC110; CPRF; KDM1; KDM1A; KIAA0601; LSD1
UniProt ID: (Human) Q8NDP6
Entrez Gene ID: (Human) 23028
Molecular Function: DNA binding protein DNA methyltransferase methyltransferase nucleic acid binding oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.