Invitrogen
Manufacturer Code:PA5114402
Catalog # PIPA5114402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human LRTOMT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Catechol O-methyltransferase 2; Catechol O-methyltransferase 2 CFAP111 COMT2 DFNB63 leucine rich transmembrane and 0-methyltransferase domain containing leucine rich transmembrane and O-methyltransferase domain containing Leucine-rich repeat-containing protein 51 leucine-rich repeat-containing protein 51 transmembrane O-methyltransferase LRRC51 LRTOMT PP7517 Protein LRTOMT1 Protein LRTOMT2 TOMT Transmembrane O-methyltransferase; catechol O-methyltransferase 2 {ECO:0000250|UniProtKB:A1Y9I9}; leucine rich transmembrane and 0-methyltransferase domain containing; Leucine-rich repeat-containing protein 51; leucine-rich repeat-containing protein 51 {ECO:0000312|RGD:1565856}; lrrc51 {ECO:0000312|RGD:1565856}; O-methyltransferase domain containing; Protein LRTOMT1; Protein LRTOMT2; tomt {ECO:0000312|RGD:1561509}; Transmembrane O-methyltransferase; Transmembrane O-methyltransferase homolog; transmembrane O-methyltransferase {ECO:0000312|RGD:1561509}
Gene Aliases: 1700008D07Rik; CFAP111; COMT2; DFNB63; LRRC51; LRTOMT; Lrtomt1; PP7517; RGD1561509; RGD1565856; TOMT
UniProt ID: (Human) Q8WZ04, (Mouse) Q9DAK8, (Rat) B6CZ61, (Rat) B6CZ62
Entrez Gene ID: (Human) 220074, (Mouse) 69358, (Rat) 293156, (Rat) 308868
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.