Abnova
Manufacturer Code:PAB24142
Catalog # 89-125-849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human LRTOMT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Protein Aliases: Catechol O-methyltransferase 2; leucine rich transmembrane and 0-methyltransferase domain containing; Leucine-rich repeat-containing protein 51; Protein LRTOMT1; Protein LRTOMT2; Transmembrane O-methyltransferase
Gene Aliases: CFAP111; COMT2; DFNB63; LRRC51; LRTOMT; PP7517; TOMT
UniProt ID: (Human) Q8WZ04
Entrez Gene ID: (Human) 220074
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.