Invitrogen
Manufacturer Code:PA543452
Catalog # PIPA543452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human LRRTM4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Leucine rich repeat Transmembrane neuronal 4 FLJ12568 MGC120633 MGC120636; leucine-rich repeat transmembrane neuronal 4 protein; Leucine-rich repeat transmembrane neuronal protein 4
Gene Aliases: 7530419J18Rik; A230052N11; LRRTM4; RGD1560707; UNQ3075/PRO9907
UniProt ID: (Human) Q86VH4, (Rat) B4F7C5, (Mouse) Q80XG9
Entrez Gene ID: (Dog) 483091, (Human) 80059, (Rabbit) 100346351, (Guinea pig) 100733877, (Rat) 500219, (Mouse) 243499, (Zebrafish) 100333709
Molecular Function:
cytokine
extracellular matrix protein
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.