Invitrogen
Manufacturer Code:PA5113080
Catalog # PIPA5113080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Leucine-rich repeat transmembrane neuronal protein 4 protein (259-419AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 7530419J18Rik A230052N11 FLJ12568 leucine rich repeat transmembrane neuronal 4 leucine-rich repeat transmembrane neuronal 4 protein Leucine-rich repeat transmembrane neuronal protein 4 Lrrtm4 MGC120633 MGC120636 RGD1560707 UNQ3075/PRO9907; leucine-rich repeat transmembrane neuronal 4 protein; Leucine-rich repeat transmembrane neuronal protein 4
Gene Aliases: 7530419J18Rik; A230052N11; LRRTM4; UNQ3075/PRO9907
UniProt ID: (Human) Q86VH4, (Mouse) Q80XG9
Entrez Gene ID: (Human) 80059, (Mouse) 243499
Molecular Function:
cytokine
extracellular matrix protein
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.