Novus Biologicals
Manufacturer Code:NBP16261120UL
Catalog # NBP162620UL
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to LRRTM4(leucine rich repeat transmembrane neuronal 4) The peptide sequence was selected form the middle region of LRRTM4. Peptide sequence FYWLKNFKGNKESTMICAGPKHIQGEKVSDAVETYNICSEVQVVNTERSH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ12568 leucine rich repeat transmembrane neuronal 4 leucine-rich repeat transmembrane neuronal 4 protein leucine-rich repeat transmembrane neuronal protein 4 MGC120633 MGC120636; leucine-rich repeat transmembrane neuronal 4 protein; Leucine-rich repeat transmembrane neuronal protein 4
Gene Aliases: LRRTM4; UNQ3075/PRO9907
UniProt ID: (Human) Q86VH4
Entrez Gene ID: (Human) 80059
Molecular Function:
cytokine
extracellular matrix protein
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.