Novus Biologicals
Manufacturer Code:NBP16006220UL
Catalog # NBP16006220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to LRRC8E(leucine rich repeat containing 8 family member E) The peptide sequence was selected from the middle region of LRRC8E. Peptide sequence LRELKQLKVLSLRSNAGKVPASVTDVAGHLQRLSLHNDGARLVALNSLKK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ23420 leucine rich repeat containing 8 family member E leucine-rich repeat-containing protein 8E; leucine-rich repeat containing 8 family member E; Leucine-rich repeat-containing protein 8E; Volume-regulated anion channel subunit LRRC8E
Gene Aliases: LRRC8E
UniProt ID: (Human) Q6NSJ5
Entrez Gene ID: (Human) 80131
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.