Invitrogen
Manufacturer Code:PA572870
Catalog # PIPA572870
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LRRC8A antibody for a 15 amino acid peptide near the amino terminus of human LRRC8A. The immunogen is within amino acids 60 - 110 of LRRC8A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ebo; HsLRRC8A; leucine rich repeat containing 8 family, member A; leucine rich repeat containing 8A; leucine-rich repeat containing 8 family member A; leucine-rich repeat-containing 8; Leucine-rich repeat-containing protein 8A; Protein ebouriffe; Swelling protein 1; Volume-regulated anion channel subunit LRRC8A; Volume-regulated anion channel subunit LRRC8A Leucine-rich repeat-containing protein 8A Swelling protein 1 LRRC8A KIAA1437 LRRC8 SWELL1
Gene Aliases: AGM5; KIAA1437; LRRC8; LRRC8A; mKIAA1437; SWELL1; UNQ221/PRO247
UniProt ID: (Human) Q8IWT6, (Mouse) Q80WG5, (Rat) Q4V8I7
Entrez Gene ID: (Human) 56262, (Mouse) 241296, (Rat) 311846
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.