Novus Biologicals
Manufacturer Code:NBP15953820UL
Catalog # NBP15953820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to LRRC8A(leucine rich repeat containing 8 family member A) The peptide sequence was selected from the middle region of LRRC8A. Peptide sequence NLTQIELRGNRLECLPVELGECPLLKRSGLVVEEDLFNTLPPEVKERLWR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGM5 FLJ10337 FLJ41617 KIAA1437leucine rich repeat containing 8 leucine rich repeat containing 8 family member A leucine-rich repeat-containing protein 8A LRRC8; HsLRRC8A; leucine rich repeat containing 8 family, member A; leucine-rich repeat containing 8 family member A; Leucine-rich repeat-containing protein 8A; Swelling protein 1; Volume-regulated anion channel subunit LRRC8A
Gene Aliases: AGM5; KIAA1437; LRRC8; LRRC8A; SWELL1; UNQ221/PRO247
UniProt ID: (Human) Q8IWT6
Entrez Gene ID: (Human) 56262
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.