OriGene
Manufacturer Code:CF805971
Catalog # 50-166-7638
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E4 |
Immunogen | Full length human recombit protein of human LRRC6 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CILD19 LRTP TSLRP; Leucine-rich repeat-containing protein 6; Leucine-rich testis-specific protein; Protein tilB homolog; Testis-specific leucine-rich repeat protein
Gene Aliases: CILD19; LRRC6; LRTP; TSLRP
UniProt ID: (Human) Q86X45
Entrez Gene ID: (Human) 23639
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.