OriGene
Manufacturer Code:CF504574
Catalog # 50-166-2985
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F9 |
Immunogen | Full length human recombit protein of human LRRC50 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CILD13 LRRC50 ODA7 swt; dynein (axonemal) assembly factor 1; Dynein assembly factor 1, axonemal; dynein, axonemal, assembly factor 1; leucine rich repeat containing 50; Leucine-rich repeat-containing protein 50; outer row dynein assembly 7 homolog; testicular tissue protein Li 110
Gene Aliases: 4930457P18Rik; CILD13; DNAAF1; LRRC50; ODA7; RGD1310542
UniProt ID: (Human) Q8NEP3, (Mouse) Q9D2H9, (Rat) Q6AYH9
Entrez Gene ID: (Human) 123872, (Mouse) 68270, (Rat) 361419
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.