OriGene
Manufacturer Code:TA504536
Catalog # 50-167-5200
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B10 |
Immunogen | Full length human recombit protein of human LRRC50 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CILD13 LRRC50 ODA7 swt; dynein (axonemal) assembly factor 1; Dynein assembly factor 1, axonemal; dynein, axonemal, assembly factor 1; leucine rich repeat containing 50; Leucine-rich repeat-containing protein 50; outer row dynein assembly 7 homolog; testicular tissue protein Li 110
Gene Aliases: CILD13; DNAAF1; LRRC50; ODA7
UniProt ID: (Human) Q8NEP3
Entrez Gene ID: (Dog) 479628, (Human) 123872
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.